Order Entry
Slovenia
ContactUsLinkComponent
 
Personal radiation detector GAMMARAE II R
  RAES047-0501-000
Personal radiation detector GAMMARAE II R
  RAES047-0501-000

 

  • Temperature range:
    −20…50 °C
  • Description:
    GammaRAE II R standard kit
  • Model:
    GammaRAE II R
  • Battery life:
    up to 600 hours
  • Dose range:
    1 µR - 999,9 R
  • Energy:
    0,06 - 3,0 MeV
  • Alarm:
    Audible, vibration and LED - can be set for DOSE or DOSE RATE
  • Datalog:
    30 000 data points ( 20 days at 60-second intervals), programmable
  • Display:
    Graphic LCD (30,5×19 mm) Radiation intensity displayed in cps or dosage rate in units of R/h or Sv/h
  • Humidity range (RH):
    0 - 95 %
  • Response time:
    < 2 seconds to alarm
  • Weight:
    270 g
  • W×D×H:
    125×68×35 mm
  • Battery type:
    2× AA batteries
  • Pk:
    1

 

 

The GammaRAE II R provides gamma detection and measurement in a rugged, portable unit. Designed for security personnel and first responders, it is intrinsically safe and features a highly sensitive scintillator sensor to detect low-level radiation and alert personnel before exposure poses a threat.

  • Fast response time to detect hidden radiation threats
  • Real-time measurement of dose and dose rate
  • Search alarm for immediate alert of a radiation threat
  • Stay-time alarm to alert user of the amount of time they can safely stay in a hazardous area
  • Continuous reading in rem/h, Sv/h, and R/h and data-logging with 30000-point capacity
  • Audible, visual and vibration alarm alerts with high and low safety alarm thresholds for both accumulated dose and dose rate
  • Bluetooth® connectivity for datalog downloading and real-time monitoring using ProRAE Guardian software

Dose Equivalent Rate (DER): 1 μR/h to 600 R/h (0,01 μSv/h to 6 Sv/h).

Complies with IEEE ANSI N42.20 standard.

 : Complies with IEEE ANSI N42.20 standard.
 : GammaRae II R kit comes with belt clip, 2 AA alkaline batteries, wrist strap, ProRAE Studio Radiation software, calibration certificate and user guide.