Order Entry
Slovenia
ContactUsLinkComponent
Anti-C19orf38 Rabbit Polyclonal Antibody
Anti-C19orf38 Rabbit Polyclonal Antibody
Catalog # BOSSBS-9065R
Supplier:  Bioss
Anti-C19orf38 Rabbit Polyclonal Antibody
Catalog # BOSSBS-9065R
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    C19orf38
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Antigen synonyms:
    Hide1|Chromosome 19 open reading frame 38|Protein HIDE1.|HIDE1_HUMAN|AB124611|C19orf38
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Concentration:
    1 ug/ul
  • Shipping temperature:
    4 °C
  • Purification:
    Purified by Protein A.
  • Size:
    100 μg
  • Pk:
    100 µl

Specifications

About this item

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: C19orf38
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human