Order Entry
Slovenia
ContactUsLinkComponent
Anti-A1CF Rabbit Polyclonal Antibody
Anti-A1CF Rabbit Polyclonal Antibody
Catalog # BOSSBS-6773R
Supplier:  Bioss
Anti-A1CF Rabbit Polyclonal Antibody
Catalog # BOSSBS-6773R
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    APOBEC1 complementation factor
  • Antigen symbol:
    A1CF
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Gene ID:
    29974
  • Antigen synonyms:
    ACF|ACF64|ACF65|ASP|APOBEC1CF
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Concentration:
    1 ug/ul
  • Shipping temperature:
    4 °C
  • Purification:
    Purified by Protein A.
  • Size:
    100 μg
  • Pk:
    100 µl

Specifications

About this item

Essential component of the apolipoprotein B mRNA editing enzyme complex which is responsible for the postranscriptional editing of a CAA codon for Gln to a UAA codon for stop in APOB mRNA. Binds to APOB mRNA and is probably responsible for docking the catalytic subunit, APOBEC1, to the mRNA to allow it to deaminate its target cytosine. The complex also protects the edited APOB mRNA from nonsense-mediated decay.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: A1CF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat