Order Entry
Slovenia
ContactUsLinkComponent
Anti-RPS6KA1 Rabbit Polyclonal Antibody
Anti-RPS6KA1 Rabbit Polyclonal Antibody
Catalog # BOSSBS-5679R
Supplier:  Bioss
Anti-RPS6KA1 Rabbit Polyclonal Antibody
Catalog # BOSSBS-5679R
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Ribosomal Protein S6 Kinase, 90kDa, Polypeptide 1
  • Antigen symbol:
    RPS6KA1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Gene ID:
    6195
  • Antigen synonyms:
    S6K-alpha|MAPKAPK-1c|p90RSK1|MAPK-activated protein kinase 1a|MAPKAPK1C|MAPK-activated protein kinase 1c|MRX19|MAP kinase-activated protein kinase 1c|HU-2|MAP kinase-activated protein kinase 1a|ISPK-1|ribosomal S6 kinase 3|MAP kinase-activated protein kinase 1b|MAPKAP kinase 1c|MAPKAP kinase 1a|r|S6K-alpha2|HU-3|90 kDa ribosomal protein S6 kinase 2|RSK-3|p90-RSK2|ribosomal protein S6 kinase alpha 2|MAPKAPK1A|pp90RSK3|RP11-393H10.3|S6K-alpha 1|RSK1|CLS|S6K-alpha-1|p90RSK2|MAPKAPK-1a|p90-RSK 2|p90-RSK 1|dJ590P13.1 (ribosomal protein S6 kinase|90kD|p90-RSK3|RSK3|HU-1|RSK|90 kDa ribosomal protein S6 kinase 1|p90S6K|ribosomal protein S6 kin|polypeptide 1)|S6K-alpha-2|RSK2|pp90RSK2|MAPKAPK1B|90 kDa ribosomal protein S6 kinase 3|S6K-alpha3|RSK-1
  • Modification:
    Thr359
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Concentration:
    1 ug/ul
  • Shipping temperature:
    4 °C
  • Purification:
    Purified by Protein A.
  • Size:
    100 μg
  • Pk:
    100 µl

Specifications

About this item

Rsk1 is a member of a family of 90kDa ribosomal protein S6 kinases, which includes Rsk1, Rsk2 and Rsk3. These are broadly expressed serine/threonine protein kinases activated in response to mitogenic stimuli, including extracellular signal regulated protein kinases Erk1 and Erk2. Rsk1 is activated by MAPK in vitro and in vivo via phosphorylation. Active Rsks appear to play a major role in transcriptional regulation by translocating to the nucleus and phosphorylating c Fos and CREB.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: RPS6KA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat