Order Entry
Slovenia
ContactUsLinkComponent
Anti-5HT3B Rabbit Polyclonal Antibody
Anti-5HT3B Rabbit Polyclonal Antibody
  BOSSBS-4289R
 :  Bioss
undefined
Anti-5HT3B Rabbit Polyclonal Antibody
  BOSSBS-4289R
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    5-hydroxytryptamine receptor 3B
  • Antigen symbol:
    5HT3B
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Gene ID:
    9177
  • Antigen synonyms:
    Serotonin receptor 3B|HTR3B|5-HT3B|5-HT3-B|5-hydroxytryptamine receptor 3B
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Concentration:
    1 ug/ul
  • Shipping temperature:
    4 °C
  • Immunogen:
    130-180/441
  • Purification:
    Purified by Protein A.
  • Size:
    100 μg
  • Pk:
    100 µl

 

 

This is one of the several different receptors for 5-hydroxytryptamine (serotonin), a biogenic hormone that functions as a neurotransmitter, a hormone, and a mitogen. This receptor is a ligand-gated ion channel, which when activated causes fast, depolarizing responses. It is a cation-specific, but otherwise relatively nonselective, ion channel.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: 5HT3B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat