Order Entry
Slovenia
ContactUsLinkComponent
Anti-UGT2B4 Rabbit Polyclonal Antibody
Anti-UGT2B4 Rabbit Polyclonal Antibody
Catalog # BOSSBS-4225R
Supplier:  Bioss
Anti-UGT2B4 Rabbit Polyclonal Antibody
Catalog # BOSSBS-4225R
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    UDP glucuronosyltransferase 2 family, polypeptide B4
  • Antigen symbol:
    UGT2B4
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Gene ID:
    7363
  • Antigen synonyms:
    UGT2B11|UDPGTH1|UDPGT2B4|HLUG25|UDPGTh-1
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Concentration:
    1 ug/ul
  • Shipping temperature:
    4 °C
  • Purification:
    Purified by Protein A.
  • Size:
    100 μg
  • Pk:
    100 µl

Specifications

About this item

The UDP-Glucuronosyltransferases (UGT) comprise a family of enzymes that detoxify and enhance the urinary excretion of a wide variety of xenobiotic and endogenous substrates by transferring glucuronic acid to sulfhydryl, hydroxyl, aromatic amino, or carboxylic acid groups. They have been subdivided into two families, UGT1 and UGT2, based on the evolutionary divergence of their genes. The enzymes of the UGT1A family play an important role in the metabolism of dietary constituents, phenols, and therapeutic drugs, and also the glucuronidation of bilirubin and iodothyronines. The enzymes of the UGT2B family are involved in the metabolism of bile acids, phenol derivatives, catecholestrogens and steroids. Although it is widely recognized that the liver is the major site of glucuronidation, it is now clear that UGT enzymes are also found in extra-hepatic tissues.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: UGT2B4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human