Anti-GPR64 Rabbit Polyclonal Antibody
Catalog # BOSSBS-12268R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:G Protein-Coupled Receptor 64
- Antigen symbol:GPR64
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Antigen synonyms:HE6|GPCR GPR64.|MGC138738|GPR64_HUMAN|Epididymis specic protein 6|G protein coupled receptor 64|GPR64|HE 6|G protein coupled receptor epididymis specic|He 6 receptor|TM7LN2|He6 receptor|MGC138739|MGC104454|GPR 64|FLJ00282
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
This orphan B G-protein coupled receptor could be involved in a signal transduction pathway controlling epidymal function and male fertility: it has been reported in the epididymis. ESTs have been isolated from embryo, kidney, placenta, skeletal muscle and testis libraries.
WB(1:100-1000), FCM(1:20-100), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: GPR64
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human