Indicating moisture traps
: Restek
Moisture trap reduces water to less than 10 ppb. Indicator changes from yellowish green to blue at 5% relative humidity. A 10 μm frit prevents microparticulate damage to needle valves and flow controllers.
- Prepurged for fast stabilisation
- Reduces noise from high sensitivity detectors
- Heavy walled glass body prevents oxygen and water infusion
Maximum operating pressure: 6,8 bar
Maximum flow: 1 L/min
Dimensions: 330×51 mm
Frequently Bought Together