Dual Vespel® ring inlet seals
Supplier: Restek
New Product
Certificates
About this item
A ring inlet seal greatly improves injection port performance as it stays sealed even after repeated temperature cycles, without retightening the reducing nut. This seal features two soft Vespel® rings, one embedded in its top surface and the other embedded in its bottom surface. These rings eliminate the need for a washer and ensure very little torque is needed to make a leak-tight seal.
Frequently Bought Together