KAPA™ stranded RNA-Seq kits with RiboErase
Supplier: KAPA BIOSYSTEMS
Certificates
About this item
The KAPA™ Stranded RNA-Seq Kit with RiboErase offers a high quality, comprehensive solution for transcriptome sequencing. By utilising a targeted enzymatic method for depletion, the workflow enables superior reduction of ribosomal RNA (rRNA) and a more complete representation of the transcriptome, including precursor mRNAs and non-coding RNA (ncRNA). Kits also contain KAPAHiFi™ for high-efficiency and low-bias library amplification, and include a streamlined, 'with-bead' protocol.
- Up to 99,98% rRNA depletion
- Flexible input of 100 ng to 1 μg total RNA from human, mouse, or rat species
- Robust and reproducible across various input amounts
- Automation-friendly workflow
Frequently Bought Together