Order Entry
Slovenia
Orders LinkContactUsLinkComponent
Counting chambers, Cellometer®
Counting chambers, Cellometer®
Supplier:   NEXCELOM BIOSCIENCE
Counting chambers, Cellometer®
Certificates
Counting chambers, Cellometer®
Counting chambers, Cellometer®
1 Options Available Below
Product Details & Documents
These counting chambers can be inserted into a cell counter, so that the cells are imaged directly.
  • Two independent enclosed chambers
  • Compatible with all Cellometer® cell counters
  • Cell suspension (20 μl) loaded via a a single channel pipette
Product Family Options
Product Information
  • Availability
    Price
  • 734-2886
    Counting chambers with protective film
    Counting chambers with protective film
    Log in for availability
    Log in for availability
    1 * 75 items
    1 * 75 items - 898,00 €
    898,00 €
    1 * 75 items
       898,00 €
  • NEXCCHT4-SD100-314
    Counting chambers with protective film
    Counting chambers with protective film
    Discontinued