Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Pumps
HPLC/GPC Pumps
Accessories for 1290 infinity LC pumps
Accessories for 1290 infinity LC pumps
Accessories for 1290 infinity LC pumps
Catalog # HEWL5067-4697
Supplier:
Agilent
Expand image
Accessories for 1290 infinity LC pumps
Catalog # HEWL5067-4697
Supplier:
Agilent
Specifications
Description:
Solvent selection valve bridge tube
Pk:
1
Specifications
About this item
Product Details
Product Details