Order Entry
Slovenia
ContactUsLinkComponent
 
Anti-ZNF585A Mouse Monoclonal Antibody [clone: 4B8]
  ABNOH00199704-M01
 :  Abnova
Anti-ZNF585A Mouse Monoclonal Antibody [clone: 4B8]
  ABNOH00199704-M01
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Zinc Finger Protein 585A
  • Antigen symbol:
    ZNF585A
  • Clonality:
    Monoclonal
  • Clone:
    4B8
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    199704
  • Storage buffer:
    1x PBS, pH 7,4
  • Sequence:
    MPANWTSPQKSSALAPEDHGSSYEGSVSFRDVAIDFSREEWRHLDPSQRNLYRDVMLETYSHLLSIGYQVPEAEVVMLEQGKEPWALQGERPRQSCPAPCLVNSHHLQESFRG
  • Storage temperature:
    Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    ZNF585A recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
  • Size:
    100 μg
  • Pk:
    100 µg

 

 

Mouse monoclonal antibody raised against a recombinant ZNF585A.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: ZNF585A
Clonality: Monoclonal
Clone: 4B8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: