Anti-FKBP4 Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: Hi52C]
ABNOMAB11388
: Abnova
- Antibody type:Primary
- Antigen name:FK506 binding protein 4
- Antigen symbol:FKBP4
- Clonality:Monoclonal
- Clone:Hi52C
- Conjugation:RPE (R-Phycoerythrin)
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Hamster,Human,Dog,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:2288
- Antigen synonyms:p52|FKBP52|Hsp56|FKBP59|FKBP51|HBI|PPIase
- Storage buffer:PBS, pH 7,4 (50% glycerol, 0,09% sodium azide)
- Storage temperature:Store at 4 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to residues of human FKBP4.
- Purification:Protein G purification
- Size:100 µg
- Pk:100 µg
Mouse monoclonal antibody raised against synthetic peptide of FKBP4.
Recommended Dilutions: Western Blot: 1:2000; Immunohistochemistry: 1:250; Immunoprecipitation: 5 ?g with 10-20 ?l Protein A beads
Type: Primary
Antigen: FKBP4
Clonality: Monoclonal
Clone: Hi52C
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Dog, Hamster, Human, Mouse, Rat