Anti-WIPI2 Rabbit Polyclonal Antibody
Catalog # ABNOPAB25670
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:WD Repeat Domain, phosphoinositide interacting 2
- Antigen symbol:WIPI2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:26100
- Antigen synonyms:Atg21|WIPI-2|CGI-50|ATG18B|WIPI49-like protein 2|WD repeat domain phosphoinositide-interacting protein 2
- Storage buffer:PBS (0,05% sodium azide)
- Storage temperature:Store at 4 °C for three months. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to 17 amino acids at N-terminus of human WIPI2.
- Purification:Peptide affinity purification
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of WIPI2.
Recommended Dilutions: Western Blot: 1-2 ?g/ml
Type: Primary
Antigen: WIPI2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat