Order Entry
Slovenia
ContactUsLinkComponent
Anti-HLA-G Mouse Monoclonal Antibody [clone: MEM-G/2]
Anti-HLA-G Mouse Monoclonal Antibody [clone: MEM-G/2]
Catalog # ABNOMAB0968
Supplier:  Abnova
Anti-HLA-G Mouse Monoclonal Antibody [clone: MEM-G/2]
Catalog # ABNOMAB0968
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    HLA class I histocompatibility antigen, alpha chain G
  • Antigen symbol:
    HLA-G
  • Clonality:
    Monoclonal
  • Clone:
    MEM-G/2
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    3135
  • Antigen synonyms:
    HLA class I histocompatibility antigen|DADB-15K14.8|alpha chain G|HLA-G histocompatibility antigen|b2 microglobulin|MHC-G|HLA class I molecule|HLA G antigen|MHC class I antigen G|class I|G
  • Storage buffer:
    PBS, pH 7,4 (15 mM sodium azide)
  • Storage temperature:
    Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant protein corresponding to heavy chain region (denaturated) of human HLA-G.
  • Size:
    100 μg
  • Pk:
    100 µg

Specifications

About this item

Mouse monoclonal antibody raised against partial recombinant HLA-G.

Type: Primary
Antigen: HLA-G
Clonality: Monoclonal
Clone: MEM-G/2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human