Anti-MSR1 Mouse Monoclonal Antibody [clone: SRA-E5]
Catalog # ABNOMAB1710
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Macrophage Scavenger Receptor 1
- Antigen symbol:MSR1
- Clonality:Monoclonal
- Clone:SRA-E5
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:4481
- Antigen synonyms:MRS-A|Scvr|SR-AII|Scara1|MSR|MSR-A|SR-AI
- Storage buffer:PBS (0,1% proclin, 2.0% Block Ace)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to human MSR1.
- Size:50 μg
- Pk:50 µg
Specifications
About this item
Mouse monoclonal antibody raised against recombinant MSR1.
Recommended Dilutions: Immunohistochemistry: 10 ?g/ml; Western Blot: 2 ?g/ml
Type: Primary
Antigen: MSR1
Clonality: Monoclonal
Clone: SRA-E5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human