Order Entry
Slovenia
ContactUsLinkComponent
Anti-biogenic amine (5HT) Receptor Rabbit Polyclonal Antibody
Anti-biogenic amine (5HT) Receptor Rabbit Polyclonal Antibody
Catalog # ABNOPAB16034
Supplier:  Abnova
Anti-biogenic amine (5HT) Receptor Rabbit Polyclonal Antibody
Catalog # ABNOPAB16034
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    biogenic amine (5HT) Receptor
  • Antigen symbol:
    biogenic amine (5HT) Receptor
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Storage buffer:
    PBS, pH 7,2 (50% glycerol, 0,01% sodium azide)
  • Storage temperature:
    Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    A synthetic peptide corresponding to C-terminus of Schistosoma mansoni biogenic amine (5HT) receptor.
  • Size:
    100 μg
  • Pk:
    100 µg

Specifications

About this item

Rabbit polyclonal antibody raised against synthetic peptide of biogenic amine (5HT) receptor.

Recommended Dilutions: ELISA: 1:2000-1:5000; Western Blot: 1 ?g/ml

Type: Primary
Antigen: biogenic amine (5HT) Receptor
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: