Anti-ASPA Goat Polyclonal Antibody
ABNOPAB19728
: Abnova
- Antibody type:Primary
- Antigen name:Aspartoacylase
- Antigen symbol:ASPA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:11484
- Antigen synonyms:ASPA, ACY2, ASP, Aspartoacylase, Aminoacylase-2
- Storage buffer:Tris saline, pH 7,3 (0,5% BSA, 0,02% sodium azide)
- Sequence:C-TLSAKSIRSTLH
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to C-terminus of mouse Aspa.
- Purification:Antigen affinity purification
- Size:100 μg
- Pk:100 µg
Goat polyclonal antibody raised against synthetic peptide of Aspa.
Recommended Dilutions: ELISA: 1:8000; Western Blot: 0.01-0.03 ?g/ml
Type: Primary
Antigen: ASPA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse, Rat