About this item
The ACK2 antibody is specific for CD117, also called c-Kit, a 145 kDa cytokine receptor important in the development of hematopoietic stem cells, in oogenesis, and for functional activity of immune cells such as NK and mast cells. c-Kit binds to a ligand known as stem cell factor (SCF), or alternatively as mast cell growth factor. Ligand binding promotes the activation (dimerisation) and subsequent tyrosine kinase activity of the c-Kit receptor and triggers key survival, expansion and maturation signals during hematopoietic progenitor cell development. Conversely, shedding of extracellular domain of c-Kit receptor is reported to induce inactivation or apoptosis within these cells. The survival signaling activity of c-Kit confers a proto-oncogenic attribute to the receptor, as overexpression or mutations in this protein are associated with tumor development.
The ACK2 antibody is widely utilised as a marker to identify hematopoietic progenitors, and to neutralise receptor-ligand binding in vitro and in vivo. Please choose the appropriate format for each application. In addition, the antibody is reported to be cross-reactive with rat c-Kit and is extensively published for use in this species.
Type: Primary
Antigen: CD117 (c-Kit)
Clonality: Monoclonal
Clone: ACK2
Conjugation: PE (Phycoerythrin)-Cy7
Epitope:
Host: Rat
Isotype: IgG2b, kappa
Reactivity: Mouse, Rat
Specifications
- Catalog No:
- 60-1172-U025
- 60-1172-U100
- Antigen symbol:
- KIT
- KIT
- Antigen name:
- Tyrosine-protein kinase Kit
- Tyrosine-protein kinase Kit
- Antigen synonyms:
- Fdc|SOW3|CD117|SCO1|Gsfsow3|W|Ssm|Tr-kit|SCO5|Gsfsco5|Gsfsco1|Bs|c-KIT
- Fdc|SOW3|CD117|SCO1|Gsfsow3|W|Ssm|Tr-kit|SCO5|Gsfsco5|Gsfsco1|Bs|c-KIT
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PE (Phycoerythrin)-Cy7®
- PE (Phycoerythrin)-Cy7®
- Clone:
- ACK2
- ACK2
- Reactivity:
- Rat, Mouse
- Rat, Mouse
- Host:
- Rat
- Rat
- Gene ID:
- 16590
- 16590
- Isotype:
- IgG2b kappa
- IgG2b kappa
- Flow cytometry:
- Yes
- Yes
- Storage temperature:
- +2 °C to +8 °C, protected from light
- +2 °C to +8 °C, protected from light
- Tested applications:
- Flow Cytometry
- Flow Cytometry