Anti-TSPY3 Rabbit Polyclonal Antibody
Catalog # ABGEAP17280B
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen symbol:TSPY3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Gene ID:100289087;728137
- Amino acid number:277 to 305
- Storage buffer:pH 7,4
- Molecular weight:35.101 kDa
- Sequence:KPPEEGTETSGDSQC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:1.350
- Shipping temperature:4 °C
- Immunogen:This TSPY3 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 277-305 amino acids from the C-terminal region of human TSPY3.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: TSPY3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human