Anti-CYP1A Mouse Monoclonal Antibody [clone: C10-7]
Supplier: Cayman Chemical
Certificates
About this item
Type: Primary
Antigen: CYP1A
Clonality: Monoclonal
Clone: C10-7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG3 kappa
Reactivity: Atlantic Cod, Carp, Lemon sole, mudfish, Rainbow Trout, Salmon, Turbot
Specifications
- Catalog No:
- CAYM173132-100
- CAYM173132-500
- Antigen symbol:
- CYP1A
- CYP1A
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- C10-7
- C10-7
- Reactivity:
- Lemon sole, Carp, Mudfish, Atlantic cod, Rainbow trout, Turbot, Salmon
- Lemon sole, Carp, Mudfish, Atlantic cod, Rainbow trout, Turbot, Salmon
- Host:
- Mouse
- Mouse
- Isotype:
- IgG3 kappa
- IgG3 kappa
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- rainbow trout CYP1A amino acids 277-294
- rainbow trout CYP1A amino acids 277-294
- Amino acid number:
- 277 to 294
- 277 to 294
- Purification:
- purified
- purified
- Cross adsorption:
- No
- No
- Storage buffer:
- 0,1 M sodium phosphate, pH 7,4, containing 0,15 M sodium chloride, 12.3 mM sodium azide, and 1% BSA
- 0,1 M sodium phosphate, pH 7,4, containing 0,15 M sodium chloride, 12.3 mM sodium azide, and 1% BSA
- Storage temperature:
- +4 °C
- +4 °C
- Shipping temperature:
- 4 °C
- 4 °C
Specifications
Frequently Bought Together