Order Entry
Slovenia
ContactUsLinkComponent
Anti-Vtg Rabbit Polyclonal Antibody [clone: PO-2]
Anti-Vtg Rabbit Polyclonal Antibody [clone: PO-2]
Supplier:  Cayman Chemical
Certificates
Anti-Vtg Rabbit Polyclonal Antibody [clone: PO-2]
Supplier:  Cayman Chemical

Specifications

  • Catalog No:
  • CAYM170150-500
  • CAYM170150-100
  • Antigen symbol:
  • VTG
  • VTG
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • PO-2
  • PO-2
  • Reactivity:
  • Carp, Sea bream, Atlantic cod, Rainbow trout, Flounder, Tilapia, Turbot, Brown trout, Wrasse, Atlantic salmon
  • Carp, Sea bream, Atlantic cod, Rainbow trout, Flounder, Tilapia, Turbot, Brown trout, Wrasse, Atlantic salmon
  • Host:
  • Rabbit
  • Rabbit
  • Isotype:
  • IgG
  • IgG
  • Western blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • Vtg from plasma of 17-beta-estradiol treated Sea Bream (S. aurata)
  • Vtg from plasma of 17-beta-estradiol treated Sea Bream (S. aurata)
  • Purification:
  • purified
  • purified
  • Cross adsorption:
  • No
  • No
  • Storage buffer:
  • 0,1 M sodium phosphate, pH 7,4, containing 0,15 M sodium chloride, 12.3 mM sodium azide, and 1% BSA
  • 0,1 M sodium phosphate, pH 7,4, containing 0,15 M sodium chloride, 12.3 mM sodium azide, and 1% BSA
  • Storage temperature:
  • +4 °C
  • +4 °C
  • Shipping temperature:
  • 4 °C
  • 4 °C

Specifications

Frequently Bought Together