Anti-Nrf2 Rabbit Polyclonal Antibody
Catalog # CAYM14114-1
Supplier: Cayman Chemical
Specifications
- Antibody type:Primary
- Antigen symbol:Nrf2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:No
- Amino acid number:18 to 29
- Storage buffer:TBS, pH 7,4, containing 5 mg/ml BSA
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:human Nrf2 N-terminal amino acids 18-29
- Purification:purified by affinity chromatography
- Size:1 Ea
- Pk:1
Specifications
About this item
Type: Primary
Antigen: Nrf2
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat