Anti-KCNQ1 Mouse Monoclonal Antibody [clone: S37A-10]
Catalog # CAYM13711-100
Supplier: Cayman Chemical
Specifications
- Antibody type:Primary
- Antigen symbol:KCNQ1
- Clonality:Monoclonal
- Clone:S37A-10
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Epitope:C-terminal
- Cross adsorption:No
- Amino acid number:2 to 101
- Storage buffer:PBS, pH 7,4, containing 50% glycerol and 0,09% sodium azide
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:fusion protein amino acids 2-101 of human KCNQ1
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: KCNQ1
Clonality: Monoclonal
Clone: S37A-10
Conjugation: unconjugated
Epitope: C-terminal
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse, Rat