Anti-DYKDDDDK Mouse Monoclonal Antibody (SureLight® APC) [clone: M2]
Supplier: Cayman Chemical
Certificates
About this item
Type: Tag
Antigen: DYKDDDDK
Clonality: Monoclonal
Clone: M2
Conjugation: SureLight® APC
Epitope:
Host: Mouse
Isotype:
Reactivity:
Specifications
- Catalog No:
- CAYM13404-100
- CAYM13404-1
- Antigen symbol:
- DYKDDDDK
- DYKDDDDK
- Antigen name:
- DYKDDDDK tag
- DYKDDDDK tag
- Antibody type:
- Tag
- Tag
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- SureLight® APC (Allophycocyanin)
- SureLight® APC (Allophycocyanin)
- Clone:
- M2
- M2
- Host:
- Mouse
- Mouse
- ImmunoFluorescence:
- Yes
- Yes
- Sequence:
- DYKDDDDK
- DYKDDDDK
- Form:
- lyophilized
- lyophilized
- Cross adsorption:
- No
- No
- Storage temperature:
- –20 °C
- –20 °C
- Shipping temperature:
- –20 °C
- –20 °C
Specifications
Frequently Bought Together