Anti-NR2A subunit of NMDA Rabbit Polyclonal Antibody
Catalog # CAYM10608-1
Supplier: Cayman Chemical
Specifications
- Antibody type:Primary
- Antigen name:NR2A subunit of NMDA
- Antigen symbol:NR2A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:No
- Form:lyophilized
- Storage buffer:5 mM ammonium bicarbonate
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:fusion protein from the C-terminus of the NR2A subunit of rat NMDA receptor
- Purification:purified by affinity chromatography
- Size:1 Ea
- Pk:1
Specifications
About this item
Type: Primary
Antigen: NR2A subunit of NMDA
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat