Anti-COX-2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # CAYM10010096-1
Supplier: Cayman Chemical
Specifications
- Antibody type:Primary
- Antigen symbol:COX-2
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Amino acid number:570 to 598
- Storage buffer:PBS, pH 7,2, containing 0,02% sodium azide
- Sequence:DPQPTKTATINASASHSRLDDINPTVLIK
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:mouse COX-2 amino acids 570-598 (DPQPTKTATINASASHSRLDDINPTVLIK)
- Purification:purified by affinity chromatography
- Size:1 Ea
- Pk:1
Specifications
About this item
Type: Primary
Antigen: COX-2
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat