Anti-S1P3 Rabbit Polyclonal Antibody
Catalog # CAYM10006373-1
Supplier: Cayman Chemical
Specifications
- Antibody type:Primary
- Antigen symbol:S1P3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:lyophilized
- Amino acid number:12 to 25
- Storage buffer:TBS, pH 7,4, containing 5 mg/ml BSA
- Sequence:VRGNETLREHYQYV
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:human S1P3 amino acids 12-25
- Purification:purified by affinity chromatography
- Size:1 Ea
- Pk:1
Specifications
About this item
Type: Primary
Antigen: S1P3
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat