Anti-Nod2 Mouse Monoclonal Antibody [clone: 2D9]
CAYM10004942-1
- Antibody type:Primary
- Antigen name:Nucleotide binding oligomerization domain containing 2
- Antigen symbol:NOD2
- Clonality:Monoclonal
- Clone:2D9
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Amino acid number:28 to 301
- Storage buffer:PBS, pH 7,4, containing 0,02% sodium azide
- Storage temperature:+4 °C
- Shipping temperature:4 °C
- Immunogen:human recombinant Nod2 amino acids 28-301
- Purification:purified by Protein A chromatography
- Size:1 Ea
- Pk:1
Type: Primary
Antigen: Nod2
Clonality: Monoclonal
Clone: 2D9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human