Anti-Vtg Rabbit Polyclonal Antibody [clone: GA-306]
Type: Primary
Antigen: Vtg
Clonality: Polyclonal
Clone: GA-306
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: ballan Wrasse, Cod, Rainbow Trout, Sheepshead Minnow, three-spined stickleback, Tilapia, Turbot
- Catalog No:
- CAYM10004442-500
- CAYM10004442-100
- Antigen symbol:
- VTG
- VTG
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- GA-306
- GA-306
- Reactivity:
- Three-spined stickleback, Sheepshead minnow, Ballan wrasse, Rainbow trout, Cod, Tilapia, Turbot
- Three-spined stickleback, Sheepshead minnow, Ballan wrasse, Rainbow trout, Cod, Tilapia, Turbot
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Vtg from whole body homogenate of 17-beta-estradiol treated three-spined stickleback (G. aculeatus)
- Vtg from whole body homogenate of 17-beta-estradiol treated three-spined stickleback (G. aculeatus)
- Purification:
- purified by affinity chromatography
- purified by affinity chromatography
- Cross adsorption:
- No
- No
- Storage buffer:
- 0,1 M sodium phosphate, pH 7,4, containing 0,15 M sodium chloride, 12.3 mM sodium azide, and 1% BSA
- 0,1 M sodium phosphate, pH 7,4, containing 0,15 M sodium chloride, 12.3 mM sodium azide, and 1% BSA
- Storage temperature:
- +4 °C
- +4 °C
- Shipping temperature:
- 4 °C
- 4 °C
Frequently Bought Together