Anti-MGST1-L1 Mouse Monoclonal Antibody [clone: 6C6]
CAYM10004350-1
- Antibody type:Primary
- Antigen symbol:MGST1-L1
- Clonality:Monoclonal
- Clone:6C6
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:lyophilized
- Storage buffer:PBS
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:recombinant human mPGE synthase-1
- Size:1 Ea
- Pk:1
Type: Primary
Antigen: MGST1-L1
Clonality: Monoclonal
Clone: 6C6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human