Order Entry
Slovenia
ContactUsLinkComponent
Anti-Zrp Mouse Monoclonal Antibody [clone: MN-2B4]
Anti-Zrp Mouse Monoclonal Antibody [clone: MN-2B4]
Supplier:  Cayman Chemical
Certificates
Anti-Zrp Mouse Monoclonal Antibody [clone: MN-2B4]
Supplier:  Cayman Chemical

Specifications

  • Catalog No:
  • CAYM100020-500
  • CAYM100020-100
  • Antigen symbol:
  • Zrp
  • Zrp
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • MN-2B4
  • MN-2B4
  • Reactivity:
  • Arctic char, Rainbow trout, Atlantic salmon
  • Arctic char, Rainbow trout, Atlantic salmon
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Western blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • eggshell proteins (Zrp and small amounts of zona pellucida proteins) from Atlantic salmon (S. salar)
  • eggshell proteins (Zrp and small amounts of zona pellucida proteins) from Atlantic salmon (S. salar)
  • Purification:
  • purified by affinity chromatography
  • purified by affinity chromatography
  • Cross adsorption:
  • No
  • No
  • Storage buffer:
  • 0,1 M sodium phosphate, pH 7,4, containing 0,15 M sodium chloride, 1% BSA, and 12.3 mM sodium azide
  • 0,1 M sodium phosphate, pH 7,4, containing 0,15 M sodium chloride, 1% BSA, and 12.3 mM sodium azide
  • Storage temperature:
  • +4 °C
  • +4 °C
  • Shipping temperature:
  • 4 °C
  • 4 °C

Specifications

Frequently Bought Together