Anti-Vtg Mouse Monoclonal Antibody [clone: CK-4B3]
Supplier: Cayman Chemical
Certificates
About this item
Type: Primary
Antigen: Vtg
Clonality: Monoclonal
Clone: CK-4B3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: japanese medaka, killifish, Sheepshead Minnow,
Specifications
- Catalog No:
- CAYM100012-500
- CAYM100012-100
- Antigen symbol:
- VTG
- VTG
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- CK-4B3
- CK-4B3
- Reactivity:
- Sheepshead minnow, Killifish, Japanese medaka
- Sheepshead minnow, Killifish, Japanese medaka
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- lipovitellin from eggs of female Japanese medaka (O. latipes)
- lipovitellin from eggs of female Japanese medaka (O. latipes)
- Purification:
- purified by affinity chromatography
- purified by affinity chromatography
- Cross adsorption:
- No
- No
- Storage buffer:
- 0,1 M sodium phosphate, pH 7,4, containing 0,15 M sodium chloride, 12.3 mM sodium azide, and 1% BSA
- 0,1 M sodium phosphate, pH 7,4, containing 0,15 M sodium chloride, 12.3 mM sodium azide, and 1% BSA
- Storage temperature:
- +4 °C
- +4 °C
- Shipping temperature:
- 4 °C
- 4 °C
Specifications
Frequently Bought Together