Anti-PGMB Goat Polyclonal Antibody
ROCK200-101-237S
- Antibody type:Primary
- Antigen symbol:PGMB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Lactococcus lacti
- Western blot:Yes
- Cross adsorption:No
- Gene ID:1114041
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide
- Storage temperature:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:Beta Phosphoglucomutase [Lactococcus lacti]
- Purification:purified by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer
- Size:25 µl
- Pk:25 µl
BETA-PHOSPHOGLUCOMUTASE Antibody has been assayed against 1.0 µg of b-Phosphoglucomutase [Lactococcus lacti] in a standard ELISA using Peroxidase conjugated Affinity Purified anti-Goat IgG [H&L] (Rabbit) and ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid])
Recommended Dilutions: ELISA: 1:5000 - 1:25000; Western Blot: 1:500 - 1:3000; Other Dilution: User Optimized
Type: Primary
Antigen: PGMB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Lactococcus lacti