Anti-CHK1 Rabbit Polyclonal Antibody
Catalog # ROCK600-401-906
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:Checkpoint Kinase 1
- Antigen symbol:CHK1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Yeast,S. cerevisiae
- Western blot:Yes
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:852577
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- Storage temperature:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:This affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding aa 312-327 of S. cerevisiae CHK1.
- Purification:affinity-purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
This affinity purified antibody has been tested for use in ELISA and by western blot. Specific conditions for reactivity should be optimized by the end user. Expect a predominant band approximately 40-60 kDa in size corresponding to CHK1 by western blotting.
Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:200 - 1:2000; Immunoprecipitation: 1:100;
Type: Primary
Antigen: CHK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Yeast, S. cerevisiae