Anti-Shc, p66 Mouse Monoclonal Antibody [clone: 6D757]
Catalog # USBIS1010-90C2-100
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:Shc, p66
- Clonality:Monoclonal
- Clone:6D757
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human,Dog,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Storage buffer:PBS, pH 7,4, 0,09% sodium azide, PEG, sucrose, 50% glycerol.
- Storage temperature:–20 °C
- Concentration:~0,1 mg/ml
- Immunogen:Synthetic peptide corresponding to a portion of N-terminal p66Shc.
- Purification:purified by thiophilic adsorption and size exclusion chromatography
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Western Blot: 0.5 ?g/ml
Type: Primary
Antigen: Shc, p66
Clonality: Monoclonal
Clone: 6D757
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Mouse, Rat,