Anti-SOD1 Mouse Monoclonal Antibody [clone: 210.29]
Catalog # AGRIAS132645
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen symbol:SOD1
- Clonality:Monoclonal
- Clone:210.29
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Amino acid number:80 to 96
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:KLH-conjugated synthetic peptide derived from human SOD1 sequence, amino acids 80-96 P00441Peptide used to elicit this antibody is not conserved in SOD2, 3 and 4.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: SOD1
Clonality: Monoclonal
Clone: 210.29
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human