Anti-MPK6 Rabbit Polyclonal Antibody
AGRIAS122633
: Agrisera
- Antibody type:Primary
- Antigen symbol:MPK6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Sheep,Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- UniProtKB:Q39026
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:KLH-conjugated synthetic peptide derived from Arabidopsis thaliana MPK6 protein,
- Size:50 µg
- Pk:50 µG
Type: Primary
Antigen: MPK6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Sheep