Anti-CrPDAT1 Rabbit Polyclonal Antibody
Catalog # AGRIAS121875
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen symbol:CRPDAT1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:recombinant CrPDAT1 without transmembrane domains, overexpressed in E.coli,
- Size:350 µl
- Pk:350 µl
Specifications
About this item
Type: Primary
Antigen: CRPDAT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human