Order Entry
Slovenia
ContactUsLinkComponent
Anti-DGAT2A Rabbit Polyclonal Antibody
Anti-DGAT2A Rabbit Polyclonal Antibody
Catalog # AGRIAS121874
Supplier:  Agrisera
undefined
Anti-DGAT2A Rabbit Polyclonal Antibody
Catalog # AGRIAS121874
Supplier:  Agrisera

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    DGAT2A
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Pinus sylvestris,
    Betula sp.,
    Picea obovata,
    Pinus strobus
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • UniProtKB:
    A8JGY1
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ambient
  • Immunogen:
    recombinant CrDGAT2A, overexpressed in E.coli, missing transmembrane domains, PID 536226, also annotated as DGTT1
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Type: Primary
Antigen: DGAT2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Betula sp., Picea obovata, Pinus strobus, Pinus sylvestris