Anti-Sub1C Rabbit Polyclonal Antibody
Catalog # AGRIAS111770
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen symbol:SUB1C
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Oryza sativa
- Western blot:Yes
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:KLH-conjugated synthetic peptide derived from Oryza sativa Sub1C. Chosen peptide is convered in all 6 isoforms (C-1 to C-6).
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: SUB1C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Oryza sativa