Anti-ADGP Rabbit Polyclonal Antibody
Catalog # AGRIAS111739
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen symbol:ADGP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:part of Arabidopsis thaliana recombinant ADP-glucose pyrophosphorylase At5g48300, P55228
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: ADGP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human