Order Entry
Slovenia
ContactUsLinkComponent
 
Anti-STN8 kinase Rabbit Polyclonal Antibody
  AGRIAS101601
 :  Agrisera
undefined
Anti-STN8 kinase Rabbit Polyclonal Antibody
  AGRIAS101601
 :  Agrisera

 

  • Antibody type:
    Primary
  • Antigen name:
    STN8 Kinase
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    A. thaliana
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Amino acid number:
    425 to 438
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ambient
  • Immunogen:
    KLH-conugated synthetic peptide (amino acids 425 - 438) specific for Arabidopsis thaliana STN8 serine/threonine proteinkinase At5g01920 (Q9LZV4)
  • Size:
    100 µg
  • Pk:
    50 µG

 

 

Type: Primary
Antigen: STN8 Kinase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana