Anti-IgG (H+L) Goat Polyclonal Antibody (Biotin)
AGRIAS101425
: Agrisera
- Antibody type:Secondary
- Antigen symbol:IgG (H+L)
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Reactivity:Pig,Rabbit,Horse,Human,Bovine,Mouse
- Western blot:Yes
- Cross adsorption:No
- Storage temperature:+2°C to +8 °C
- Shipping temperature:Ambient
- Immunogen:Purified mouse IgG (H&L) AAA51107
- Purification:purified
- Size:2 mg
- Pk:2 mg
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Horse, Human, Mouse, Pig, Rabbit