Anti-kappa light chain Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # AGRIAS101412
Supplier: Agrisera
Specifications
- Antibody type:Secondary
- Antigen name:Kappa Light Chain
- Antigen symbol:IGKC
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Flow cytometry:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Cross adsorption:No
- Storage temperature:+2°C to +8 °C
- Shipping temperature:Ambient
- Immunogen:Purified human Kappa Chain BAA37169.1
- Purification:purified
- Size:500 μg
- Pk:0,5 mg
Specifications
About this item
Type: Secondary
Antigen: kappa light chain
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse