Anti-MnSOD Rabbit Polyclonal Antibody
AGRIAS09524
: Agrisera
- Antibody type:Primary
- Antigen symbol:Mn/SOD
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Mus musculus
- Western blot:Yes
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:KLH-conjugated synthetic peptide derived from available MnSOD sequences in di and monocotyl plants including Arabidopsis thaliana O81235, At3g10920
- Size:100 µl
- Pk:50 µl
Type: Primary
Antigen: Mn/SOD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mus musculus