Order Entry
Slovenia
ContactUsLinkComponent
Anti-UBQ11 Rabbit Polyclonal Antibody
Anti-UBQ11 Rabbit Polyclonal Antibody
Catalog # AGRIAS08307A
Supplier:  Agrisera
undefined
Anti-UBQ11 Rabbit Polyclonal Antibody
Catalog # AGRIAS08307A
Supplier:  Agrisera

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    UBQ11
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Oryza sativa,
    N. benthamiana,
    A. thaliana
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ambient
  • Immunogen:
    recombinant Arabidopsis thaliana ubiquitin, ubq11, with N-terminal His-tag; TAIR:  At4g05050
  • Purification:
    affinity-purified
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Type: Primary
Antigen: UBQ11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana, N. benthamiana, Oryza sativa