Anti-LOX-C Rabbit Polyclonal Antibody
Catalog # AGRIAS07258
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen symbol:LOX-C
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Lilium longiflorum,Olea europaea,A. thaliana,Soy bean,Lupinus luteus
- Western blot:Yes
- Cross adsorption:No
- Amino acid number:257 to 450
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:recombinant Arabidopsis thaliana protein, loop (aa 257-450) P38418, At3g45140
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: LOX-C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana, glycine max, Lilium longiflorum, Lupinus luteus, Olea europaea