Anti-V-ATPase Rabbit Polyclonal Antibody
Catalog # AGRIAS07213
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen name:V-ATPase
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Petunia sp.,Noccaea caerulescens,Pteris vittata,Oryza sativa,Thellungiella sp.,Lilium longiflorum,C. reinhardtii,Zea mays,A. thaliana,N. tabacum,L. esculentum,Hordeum vulgare,Mesembryanthemum sp.,A. comosus
- Western blot:Yes
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:KLH-conjugated synthetic peptide chosen from subunit E of plant V-ATPase including Arabidopsis thaliana At4g11150. Peptide is conserved in vacuolar H+-ATPase subunit E, isoform 1 to 3 (VHA-E1).
- Size:200 µl
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: V-ATPase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. comosus, A. thaliana, C. reinhardtii, Hordeum vulgare, Lilium longiflorum, Mesembryanthemum sp., N. tabacum, L. esculentum, Noccaea caerulescens, Oryza sativa, Petunia sp., Pteris vittata, Thellungiella sp., Zea mays